CAS RN 87967-95-1
IUPAC Name:
5-methyl-3-phenyl-1,2-oxazole-4-carbaldehyde
Synonyms:
5-methyl-3-phenyl-1,2-oxazole-4-carbaldehyde
Molecular Weight (g/mol):
187.2
Molecular Formula:
C11H9NO2
InChi Key:
SMSBKEHQYUYAHK-UHFFFAOYSA-N
SMILES:
CC1=C(C=O)C(=NO1)C1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
5-Methyl-3-phenylisoxazole-4-carboxaldehyde, 97%
5-Methyl-3-phenylisoxazole-4-carboxaldehyde, 97%
CAS: 87967-95-1 Molecular Formula: C11H9NO2 Molecular Weight (g/mol): 187.20 MDL Number: MFCD02677714 InChI Key: SMSBKEHQYUYAHK-UHFFFAOYSA-N Synonym: 5-methyl-3-phenyl-4-isoxazolecarbaldehyde,5-methyl-3-phenylisoxazole-4-carbaldehyde,5-methyl-3-phenylisoxazole-4-carboxaldehyde,5-methyl-3-phenyl-4-isoxazolecarboxaldehyde,5-methyl-3-phenyl-isoxazole-4-carbaldehyde,acmc-20am24,3-phenyl-5-methyl-4-isoxazolecarbaldehyde,3-phenyl-5-methylisoxazole-4-carbaldehyde,4-isoxazolecarboxaldehyde,5-methyl-3-phenyl,5-methyl-3-phenyl-4-isoxazole carboxaldehyde PubChem CID: 2776144 IUPAC Name: 5-methyl-3-phenyl-1,2-oxazole-4-carbaldehyde SMILES: CC1=C(C=O)C(=NO1)C1=CC=CC=C1