accessibility menu, dialog, popup

UserName

H3CBrN

CAS RN 877-42-9

IUPAC Name: 6-bromo-2-methylquinoline
Synonyms: 6-bromo-2-methylquinoline
Molecular Weight (g/mol): 222.09
Molecular Formula: C10H8BrN
InChi Key: SQRYQSKJZVQJAY-UHFFFAOYSA-N
SMILES: CC1=CC=C2C=C(Br)C=CC2=N1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)