missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-2-methylquinoline, 97%

Catalog No. AAH2744714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2744714 25 g
AAH2744706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2744714 Supplier Thermo Scientific Chemicals Supplier No. H2744714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 877-42-9 | C10H8BrN | 222.085 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 877-42-9
Molecular Formula C10H8BrN
Molecular Weight (g/mol) 222.085
MDL Number MFCD00079724
InChI Key SQRYQSKJZVQJAY-UHFFFAOYSA-N
Synonym 6-bromoquinaldine, 2-methyl-6-bromoquinoline, 6-bromo-2-methyl-quinoline, quinoline, 6-bromo-2-methyl, 6-bromo-2-methyl-1-azanaphthalene, zlchem 926, pubchem5882, acmc-209qps, ksc494e7l
PubChem CID 522885
IUPAC Name 6-bromo-2-methylquinoline
SMILES CC1=NC2=C(C=C1)C=C(C=C2)Br

Specifications

Melting Point 100°C to 103°C
Quantity 25 g
Formula Weight 222.08
Percent Purity 97%
Chemical Name or Material 6-Bromo-2-methylquinoline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only