accessibility menu, dialog, popup

UserName

CH3(E)

CAS RN 873-66-5

IUPAC Name: [(1E)-prop-1-en-1-yl]benzene
Synonyms: trans-1-phenyl-1-propene
Molecular Weight (g/mol): 118.18
Molecular Formula: C9H10
InChi Key: QROGIFZRVHSFLM-QHHAFSJGSA-N
SMILES: C\C=C\C1=CC=CC=C1
Boiling Point 
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)