accessibility menu, dialog, popup

UserName

CAS RN 872511-34-7

H3CNNNHNNNH3COHNClOCH3OCH3Cl

CAS RN 872511-34-7

IUPAC Name: 1-(2,6-dichloro-3,5-dimethoxyphenyl)-3-(6-{[4-(4-ethylpiperazin-1-yl)phenyl]amino}pyrimidin-4-yl)-3-methylurea
Synonyms: nvp-BGJ398
Molecular Weight (g/mol): 560.48
Molecular Formula: C26H31Cl2N7O3
InChi Key: QADPYRIHXKWUSV-UHFFFAOYSA-N
SMILES: CCN1CCN(CC1)C1=CC=C(NC2=CC(=NC=N2)N(C)C(=O)NC2=C(Cl)C(OC)=CC(OC)=C2Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results