accessibility menu, dialog, popup

UserName

CAS RN 871-84-1

IUPAC Name: octa-1,7-diyne
Synonyms: 1,7-octadiyne
Molecular Weight (g/mol): 106.17
Molecular Formula: C8H10
InChi Key: DSOJWVLXZNRKCS-UHFFFAOYSA-N
SMILES: C#CCCCCC#C
Boiling Point 
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (2)
  • (4)