missing translation for 'onlineSavingsMsg'
Learn More

1,7-Octadiyne 98.0+%, TCI America™

Catalog No. O01475ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O01475ML 5 mL
O014725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O01475ML Supplier TCI America Supplier No. O01475ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 871-84-1
Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
MDL Number MFCD00008580
InChI Key DSOJWVLXZNRKCS-UHFFFAOYSA-N
Synonym 1,7-octadiyne, 1,7 octadiyne, oct-1,7-diyne, acmc-209qii, 3-thia-1,5-pentanediamine, 1,7-octadiyne 10g, 1,7-octadiyne 6ci, 7ci, 8ci, 9ci
PubChem CID 70099
IUPAC Name octa-1,7-diyne
SMILES C#CCCCCC#C

Specifications

Color Yellow
Boiling Point 135°C
Quantity 5 mL
UN Number 3295
Formula Weight 106.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,7-Octadiyne