missing translation for 'onlineSavingsMsg'
Learn More

1,7-Octadiyne, 98%

Catalog No. AAA1582918
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1582918 50 g
AAA1582909 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1582918 Supplier Thermo Scientific Chemicals Supplier No. A1582918
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 871-84-1 | C8H10 | 106.17 g/mol

1,7-Octadiyne is an useful bifunctional chain extension unit. It participates in the formation of uranium(IV) vinyl complexes. It reacts with frustrated Lewis pair P(o-tolyl)3/B(C6F5)3 by acetylene C-C coupling to yield the zwitterionic product. It undergoes the reductive cyclization during reaction with Re2Cl4(?-dppm)2 (dppm = Ph2PCH2PPh2) and affords quadruply bonded dirhenium(III) complex Re2Cl3(?,η2-C8H7)(?-dppm)2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,7-Octadiyne is an useful bifunctional chain extension unit. It participates in the formation of uranium(IV) vinyl complexes. It reacts with frustrated Lewis pair P(o-tolyl)3/B(C6F5)3 by acetylene C-C coupling to yield the zwitterionic product. It undergoes the reductive cyclization during reaction with Re2Cl4(μ-dppm)2 (dppm = Ph2PCH2PPh2) and affords quadruply bonded dirhenium(III) complex Re2Cl3(μ,η2-C8H7)(μ-dppm)2.

Solubility
Not miscible or difficult to mix.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 871-84-1
Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
MDL Number MFCD00008580
InChI Key DSOJWVLXZNRKCS-UHFFFAOYSA-N
Synonym 1,7-octadiyne, 1,7 octadiyne, oct-1,7-diyne, acmc-209qii, 3-thia-1,5-pentanediamine, 1,7-octadiyne 10g, 1,7-octadiyne 6ci, 7ci, 8ci, 9ci
PubChem CID 70099
IUPAC Name octa-1,7-diyne
SMILES C#CCCCCC#C

Specifications

Density 0.81
Boiling Point 135°C to 136°C
Flash Point 23°C (73°F)
Odor Recognizable
Refractive Index 1.446
Quantity 50 g
UN Number UN3295
Beilstein 1697275
Solubility Information Not miscible or difficult to mix.
Formula Weight 106.17
Percent Purity 98%
Chemical Name or Material 1,7-Octadiyne
Show More Show Less
Hazard Category H226-H315-H319
Hazard Statement GHS H Statement
H304-H226-H315-H319-H335
May be fatal if swallowed and enters airways.
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 212-815-0
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only