CAS RN 87-51-4
Indole-3-acetic acid, 98+%
CAS: 87-51-4 Molecular Formula: C10H9NO2 Molecular Weight (g/mol): 175.19 MDL Number: MFCD00005636 InChI Key: SEOVTRFCIGRIMH-UHFFFAOYSA-N Synonym: indole-3-acetic acid,3-indoleacetic acid,heteroauxin,indoleacetic acid,1h-indole-3-acetic acid,2-1h-indol-3-yl acetic acid,rhizopin,1h-indol-3-ylacetic acid,indol-3-ylacetic acid,rhizopon a PubChem CID: 802 ChEBI: CHEBI:16411 IUPAC Name: 2-(1H-indol-3-yl)acetic acid SMILES: OC(=O)CC1=CNC2=CC=CC=C12
1H-Indole-3-acetic acid, 98%
CAS: 87-51-4 Molecular Formula: C10H9NO2 Molecular Weight (g/mol): 175.19 MDL Number: MFCD00005636 InChI Key: SEOVTRFCIGRIMH-UHFFFAOYSA-N Synonym: indole-3-acetic acid,3-indoleacetic acid,heteroauxin,indoleacetic acid,1h-indole-3-acetic acid,2-1h-indol-3-yl acetic acid,rhizopin,1h-indol-3-ylacetic acid,indol-3-ylacetic acid,rhizopon a PubChem CID: 802 ChEBI: CHEBI:16411 IUPAC Name: 2-(1H-indol-3-yl)acetic acid SMILES: OC(=O)CC1=CNC2=CC=CC=C12
Indole-3-acetic acid, 99.2%, MP Biomedicals™
CAS: 87-51-4 Molecular Formula: C10H9NO2 Molecular Weight (g/mol): 175.19 MDL Number: MFCD00005636 InChI Key: SEOVTRFCIGRIMH-UHFFFAOYSA-N Synonym: indole-3-acetic acid,3-indoleacetic acid,heteroauxin,indoleacetic acid,1h-indole-3-acetic acid,2-1h-indol-3-yl acetic acid,rhizopin,1h-indol-3-ylacetic acid,indol-3-ylacetic acid,rhizopon a PubChem CID: 802 ChEBI: CHEBI:16411 IUPAC Name: 2-(1H-indol-3-yl)acetic acid SMILES: OC(=O)CC1=CNC2=CC=CC=C12
3-Indoleacetic Acid 98.0+%, TCI America™
CAS: 87-51-4 Molecular Formula: C10H9NO2 Molecular Weight (g/mol): 175.19 MDL Number: MFCD00005636 InChI Key: SEOVTRFCIGRIMH-UHFFFAOYSA-N Synonym: indole-3-acetic acid,3-indoleacetic acid,heteroauxin,indoleacetic acid,1h-indole-3-acetic acid,2-1h-indol-3-yl acetic acid,rhizopin,1h-indol-3-ylacetic acid,indol-3-ylacetic acid,rhizopon a PubChem CID: 802 ChEBI: CHEBI:16411 IUPAC Name: 2-(1H-indol-3-yl)acetic acid SMILES: OC(=O)CC1=CNC2=CC=CC=C12
Tocris Bioscience™ Auxin
Chemical dimerizer used in auxin-inducible degron (AID) system; phytohormone