accessibility menu, dialog, popup

UserName

O

CAS RN 86505-44-4

IUPAC Name: 3-phenylcyclopentan-1-one
Synonyms: 3-phenylcyclopentan-1-one
Molecular Weight (g/mol): 160.22
Molecular Formula: C11H12O
InChi Key: MFBXYJLOYZMFIN-UHFFFAOYNA-N
SMILES: O=C1CCC(C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results