missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-3-Phenylcyclopentanone 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 100 mg |
| Formula Weight | 160.22 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | (R)-3-Phenylcyclopentanone |
Safety and Handling
| TSCA | No |