accessibility menu, dialog, popup

UserName

CAS RN 865-49-6

IUPAC Name: trichloro(²H)methane
Synonyms: deuterated chloroform
Molecular Weight (g/mol): 120.38
Molecular Formula: CHCl3
InChi Key: HEDRZPFGACZZDS-MICDWDOJSA-N
SMILES: [2H]C(Cl)(Cl)Cl
Concentration 
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)