missing translation for 'onlineSavingsMsg'
Learn More

Iodomethane-13C Solution NMR Reference Standard, MilliporeSigma™ Supelco™

Catalog No. 111002450
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002450 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002450 Supplier Merck Emd Millipore Supplier No. 6113361EA

Indirect detection test

Specifications

CAS 865-49-6
Concentration 1% in chloroform-d (99.9 atom% D)
Linear Formula 13 CH3 I
MDL Number MFCD00001075
Type NMR Reference Standard
Quantity 1 Ea.
Formula Weight 142.93
Grade NMR Reference Standard
Concentration or Composition (by Analyte or Components) chromium(III) acetylacetonate, 0.2%