accessibility menu, dialog, popup

UserName

CH3H3CCH3NNBr

CAS RN 85929-94-8

IUPAC Name: 5-bromo-2-tert-butylpyrimidine
Synonyms: 5-bromo-2-tert-butylpyrimidine
Molecular Weight (g/mol): 215.09
Molecular Formula: C8H11BrN2
InChi Key: FAAQGONYZKWBKC-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=NC=C(Br)C=N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results