accessibility menu, dialog, popup

UserName

ONNNClO

CAS RN 851030-18-7

IUPAC Name: 2,4-bis(benzyloxy)-6-chloro-1,3,5-triazine
Synonyms: 2,4-bis(benzyloxy)-6-chloro-1,3,5-triazine
Molecular Weight (g/mol): 327.77
Molecular Formula: C17H14ClN3O2
InChi Key: XKDFUXJROYAXAO-UHFFFAOYSA-N
SMILES: ClC1=NC(OCC2=CC=CC=C2)=NC(OCC2=CC=CC=C2)=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results