accessibility menu, dialog, popup

UserName

CAS RN 850413-36-4

CH3NNCH3Br

CAS RN 850413-36-4

IUPAC Name: 6-bromo-4,4'-dimethyl-2,2'-bipyridine
Synonyms: 6-bromo-4,4'-dimethyl-2,2'-bipyridine
Molecular Weight (g/mol): 263.14
Molecular Formula: C12H11BrN2
InChi Key: UEJJXCRFGURPDR-UHFFFAOYSA-N
SMILES: CC1=CC(=NC=C1)C1=NC(Br)=CC(C)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results