missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-4,4'-dimethyl-2,2'-bipyridyl 97.0+%, TCI America™

Catalog No. p-7123130
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
B37321G 1 g
B37325G 5 g
2 options

Catalog No. B37321G

Supplier: TCI America B37321G

Only null left

Chemical Identifiers

CAS 850413-36-4
Molecular Formula C12H11BrN2
Molecular Weight (g/mol) 263.14
MDL Number MFCD11042495
InChI Key UEJJXCRFGURPDR-UHFFFAOYSA-N
PubChem CID 53438778
IUPAC Name 6-bromo-4,4'-dimethyl-2,2'-bipyridine
SMILES CC1=CC(=NC=C1)C1=NC(Br)=CC(C)=C1

Specifications

Melting Point 107°C
Color White-Yellow
Quantity 1 g
Synonym 6-Bromo-4,4′C-dimethyl-2,2′C-bipyridine
Formula Weight 263.14
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Bromo-4,4′-dimethyl-2,2′-bipyridyl