accessibility menu, dialog, popup

UserName

CAS RN 850036-28-1

IUPAC Name: (cyclopent-1-en-1-yl)boronic acid
Synonyms: cyclopent-1-en-1-ylboronic acid
Molecular Weight (g/mol): 111.94
Molecular Formula: C5H9BO2
InChi Key: UZBHNSVUMGIKLU-UHFFFAOYSA-N
SMILES: OB(O)C1=CCCC1
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)