missing translation for 'onlineSavingsMsg'
Learn More

1-Cyclopentenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. C28931G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C28931G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C28931G Supplier TCI America Supplier No. C28931G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 850036-28-1
Molecular Formula C5H9BO2
Molecular Weight (g/mol) 111.935
MDL Number MFCD02179496
InChI Key UZBHNSVUMGIKLU-UHFFFAOYSA-N
PubChem CID 3861343
IUPAC Name cyclopenten-1-ylboronic acid
SMILES B(C1=CCCC1)(O)O

Specifications

Melting Point 140°C
Quantity 1 g
Synonym cyclopent-1-en-1-ylboronic acid, 1-cyclopentenylboronic acid, cyclopentenylboronic acid, cyclopent-1-en-1-yl boronic acid, cyclopentene-1-boronic acid, cyclopenten-1-yl boronic acid, cyclopent-1-ene-1-boronic acid, boronic acid, 1-cyclopenten-1-yl, cyclopenteneboronic acid, acmc-209pzb
Formula Weight 111.94
Physical Form Crystalline Powder
Chemical Name or Material 1-Cyclopentenylboronic Acid (contains varying amounts of Anhydride)