accessibility menu, dialog, popup

UserName

CAS RN 85-91-6

IUPAC Name: methyl 2-(methylamino)benzoate
Synonyms: methyl 2-(methylamino)benzoate
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: GVOWHGSUZUUUDR-UHFFFAOYSA-N
SMILES: CNC1=CC=CC=C1C(=O)OC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results