missing translation for 'onlineSavingsMsg'
Learn More

Methyl N-methylanthranilate, 98%

Catalog No. AAB2023330
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2023330 250 g
AAB2023318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2023330 Supplier Thermo Scientific Chemicals Supplier No. B2023330
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 85-91-6 | C9H11NO2 | 165.19 g/mol

Methyl N-methylanthranilate is a fragrance ingredient used in decorative cosmetics, fine fragrances, shampoos, toilet soaps and other toiletries as well as in non-cosmetic products such as household cleaners and detergents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl N-methylanthranilate is a fragrance ingredient used in decorative cosmetics, fine fragrances, shampoos, toilet soaps and other toiletries as well as in non-cosmetic products such as household cleaners and detergents.

Solubility
Miscible in all proportions with ethanol 96%, DMSO and diethyl ether.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 85-91-6
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00017183
InChI Key GVOWHGSUZUUUDR-UHFFFAOYSA-N
Synonym methyl 2-methylamino benzoate, methyl n-methylanthranilate, dimethyl anthranilate, methyl methylaminobenzoate, methyl methanthranilate, methyl methylanthranilate, benzoic acid, 2-methylamino-, methyl ester, methyl n-methyl anthranilate, methyl n-methyl-o-anthranilate, methyl o-methylamino benzoate
PubChem CID 6826
IUPAC Name methyl 2-(methylamino)benzoate
SMILES CNC1=CC=CC=C1C(=O)OC

Specifications

Melting Point 17°C to 19°C
Boiling Point 256°C to 258°C
Flash Point 133°C (271°F)
Quantity 250 g
Beilstein 607217
Formula Weight 165.19
Percent Purity 98%
Chemical Name or Material Methyl N-methylanthranilate
RTECSNumber CB3500000
TSCA TSCA

RUO – Research Use Only