accessibility menu, dialog, popup

UserName

CAS RN 849061-97-8

IUPAC Name: 2-(3,4-dihydroxyphenyl)-3,5,7-trihydroxy-4H-chromen-4-one
Synonyms: quercetin
Molecular Weight (g/mol): 302.24
Molecular Formula: C15H10O7
InChi Key: REFJWTPEDVJJIY-UHFFFAOYSA-N
SMILES: OC1=CC(O)=C2C(OC(=C(O)C2=O)C2=CC(O)=C(O)C=C2)=C1
Percent Purity 
  • (1)
  • (3)
Physical Form 
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)