missing translation for 'onlineSavingsMsg'
Learn More

Quercetin Hydrate 96.0+%, TCI America™

Catalog No. P004225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P004225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P004225G Supplier TCI America Supplier No. P004225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 849061-97-8
Molecular Formula C15H10O7
Molecular Weight (g/mol) 302.24
MDL Number MFCD03847906
InChI Key REFJWTPEDVJJIY-UHFFFAOYSA-N
Synonym quercetin hydrate, 2-3,4-dihydroxyphenyl-3,5,7-trihydroxy-4h-chromen-4-one hydrate, quercetinhydrate, quercetine sophoretin /, regid_for_cid_747934, regid_for_cid_16212154, 3,3;,4;,5,7-pentahydroxyflavon, 2-3,4-dihydroxyphenyl-3,5,7-trihydroxychromen-4-one, hydrate
PubChem CID 16212154
IUPAC Name 2-(3,4-dihydroxyphenyl)-3,5,7-trihydroxy-4H-chromen-4-one
SMILES OC1=CC(O)=C2C(OC(=C(O)C2=O)C2=CC(O)=C(O)C=C2)=C1

Specifications

Color Yellow
Quantity 25 g
UN Number 2811
Formula Weight 302.24
Percent Purity ≥96.0% (HPLC,E)
Physical Form Crystalline Powder
Chemical Name or Material Quercetin Hydrate
RTECSNumber LK8750000
TSCA Yes