accessibility menu, dialog, popup

UserName

CH3SiH3CCH3OSOOFFFSiH3CH3CCH3OSOOFFFSiCH3H3CCH3OSOOFFF

CAS RN 847925-63-7

IUPAC Name: 4-[4'-(trifluoromethanesulfonyloxy)-5-[4-(trifluoromethanesulfonyloxy)-3-(trimethylsilyl)phenyl]-3'-(trimethylsilyl)-[1,1'-biphenyl]-3-yl]-2-(trimethylsilyl)phenyl trifluoromethanesulfonate
Synonyms: 4-[4'-(trifluoromethanesulfonyloxy)-5-[4-(trifluoromethanesulfonyloxy)-3-(trimethylsilyl)phenyl]-3'-(trimethylsilyl)-[1,1'-biphenyl]-3-yl]-2-(trimethylsilyl)phenyl trifluoromethanesulfonate
Molecular Weight (g/mol): 967.12
Molecular Formula: C36H39F9O9S3Si3
InChi Key: ZLEUXQBUQGSSOW-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C1=C(OS(=O)(=O)C(F)(F)F)C=CC(=C1)C1=CC(=CC(=C1)C1=CC(=C(OS(=O)(=O)C(F)(F)F)C=C1)[Si](C)(C)C)C1=CC(=C(OS(=O)(=O)C(F)(F)F)C=C1)[Si](C)(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results