missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Tris[4-(trifluoromethanesulfonyloxy)-3-(trimethylsilyl)phenyl]benzene 92.0+%, TCI America™

Catalog No. T24671G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24671G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T24671G Supplier TCI America Supplier No. T24671G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 847925-63-7
Molecular Formula C36H39F9O9S3Si3
Molecular Weight (g/mol) 967.12
MDL Number MFCD10000966
InChI Key ZLEUXQBUQGSSOW-UHFFFAOYSA-N
PubChem CID 44629864
IUPAC Name 4-[4'-(trifluoromethanesulfonyloxy)-5-[4-(trifluoromethanesulfonyloxy)-3-(trimethylsilyl)phenyl]-3'-(trimethylsilyl)-[1,1'-biphenyl]-3-yl]-2-(trimethylsilyl)phenyl trifluoromethanesulfonate
SMILES C[Si](C)(C)C1=C(OS(=O)(=O)C(F)(F)F)C=CC(=C1)C1=CC(=CC(=C1)C1=CC(=C(OS(=O)(=O)C(F)(F)F)C=C1)[Si](C)(C)C)C1=CC(=C(OS(=O)(=O)C(F)(F)F)C=C1)[Si](C)(C)C

Specifications

Color White-Yellow
Quantity 1 g
UN Number 3261
Formula Weight 967.12
Percent Purity ≥92.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,5-Tris[4-(trifluoromethanesulfonyloxy)-3-(trimethylsilyl)phenyl]benzene