accessibility menu, dialog, popup

UserName

CH3(R)OO(R)H3C(S)O(R)(R)H3COH3C(S)OH(S)CH3OH3C(R)O(S)O(R)CH3(S)(R)HONCH3H3C(R)CH3HO(R)CH3NH3C(R)CH3(R)HO(S)H3COH

CAS RN 83905-01-5

IUPAC Name: (2R,3S,4R,5R,8R,10R,11R,13S,14R)-11-{[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy}-2-ethyl-3,4,10-trihydroxy-13-{[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy}-3,5,6,8,10,12,14-heptamethyl-1-oxa-6-azacyclopentadecan-15-one
Synonyms: azithromycin
Molecular Weight (g/mol): 749
Molecular Formula: C38H72N2O12
InChi Key: MQTOSJVFKKJCRP-OHJWJPDZSA-N
SMILES: CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)C(C)[C@@H](O[C@@H]2O[C@H](C)C[C@@H]([C@H]2O)N(C)C)[C@](C)(O)C[C@@H](C)CN(C)[C@H](C)[C@@H](O)[C@]1(C)O
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results