accessibility menu, dialog, popup

UserName

FFFFFF

CAS RN 83558-76-3

IUPAC Name: (1,1,1,3,3,3-hexafluoro-2-phenylpropan-2-yl)benzene
Synonyms: (1,1,1,3,3,3-hexafluoro-2-phenylpropan-2-yl)benzene
Molecular Weight (g/mol): 304.24
Molecular Formula: C15H10F6
InChi Key: CFTSORNHIUMCGF-UHFFFAOYSA-N
SMILES: FC(F)(F)C(C1=CC=CC=C1)(C1=CC=CC=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results