accessibility menu, dialog, popup

UserName

CAS RN 83-55-6

IUPAC Name: 5-aminonaphthalen-1-ol
Synonyms: 5-aminonaphthalen-1-ol
Molecular Weight (g/mol): 159.19
Molecular Formula: C10H9NO
InChi Key: ZBIBQNVRTVLOHQ-UHFFFAOYSA-N
SMILES: NC1=C2C=CC=C(O)C2=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results