accessibility menu, dialog, popup

UserName

CH3ONHSOOOOHOCH3Na

CAS RN 82692-88-4

IUPAC Name: sodium 3-[(3,5-dimethoxyphenyl)amino]-2-hydroxypropane-1-sulfonate
Synonyms: sodium 3-[(3,5-dimethoxyphenyl)amino]-2-hydroxypropane-1-sulfonate
Molecular Weight (g/mol): 313.3
Molecular Formula: C11H16NNaO6S
InChi Key: SVLRFMQGKVFRTB-UHFFFAOYNA-M
SMILES: [Na+].COC1=CC(OC)=CC(NCC(O)CS([O-])(=O)=O)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results