accessibility menu, dialog, popup

UserName

HO

CAS RN 825-51-4

IUPAC Name: decahydronaphthalen-2-ol
Synonyms: decahydro-2-naphthol
Molecular Weight (g/mol): 154.25
Molecular Formula: C10H18O
InChi Key: UPMAOXLCTXPPAG-UHFFFAOYNA-N
SMILES: OC1CCC2CCCCC2C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results