missing translation for 'onlineSavingsMsg'
Learn More

Decahydro-2-naphthol (mixture of isomers) 95.0+%, TCI America™

Catalog No. D200025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D200025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D200025G Supplier TCI America Supplier No. D200025G

Chemical Identifiers

CAS 825-51-4
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.253
MDL Number MFCD00004132
InChI Key UPMAOXLCTXPPAG-UHFFFAOYSA-N
Synonym 2-hydroxydecalin, 2-decalol, 2-naphthalenol, decahydro, decalin-2-ol, decahydronaphthalen-2-ol, 2-decalinol, decahydro-2-naphthol, decahydronapthol-2, 2-naphthol, decahydro, decahydronaphthol-2
PubChem CID 13216
IUPAC Name 1,2,3,4,4a,5,6,7,8,8a-decahydronaphthalen-2-ol
SMILES C1CCC2CC(CCC2C1)O

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 154.25
Percent Purity ≥95.0% (GC)
Physical Form Paste
Chemical Name or Material Decahydro-2-naphthol (mixture of isomers)
RTECSNumber QK4240000
TSCA Yes