accessibility menu, dialog, popup

UserName

NH2ClNOO

CAS RN 825-41-2

IUPAC Name: 3-chloro-4-nitroaniline
Synonyms: 3-chloro-4-nitroaniline
Molecular Weight (g/mol): 172.57
Molecular Formula: C6H5ClN2O2
InChi Key: LDSIOPGMLLPSSR-UHFFFAOYSA-N
SMILES: NC1=CC=C(C(Cl)=C1)[N+]([O-])=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)