missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-nitroaniline, 95%

Catalog No. AAH3258109
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3258109 10 g
AAH3258103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3258109 Supplier Thermo Scientific Chemicals Supplier No. H3258109
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 825-41-2 | C6H5ClN2O2 | 172.568 g/mol

3-Chloro-4-nitroaniline is used as a pharmaceutical and medical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chloro-4-nitroaniline is used as a pharmaceutical and medical intermediate.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 825-41-2
Molecular Formula C6H5ClN2O2
Molecular Weight (g/mol) 172.568
MDL Number MFCD00085922
InChI Key LDSIOPGMLLPSSR-UHFFFAOYSA-N
Synonym benzenamine, 3-chloro-4-nitro, aniline, 3-chloro-4-nitro, 3-chlor-4-nitroanilin, 3-chloro-4-nitrobenzenamine, zlchem 908, pubchem9594, acmc-209ppd, 2-chloro-4-aminonitrobenzene, 4-amino-2-chloronitrobenzene, 3-chloro-4-nitro-phenylamine
PubChem CID 69996
IUPAC Name 3-chloro-4-nitroaniline
SMILES C1=CC(=C(C=C1N)Cl)[N+](=O)[O-]

Specifications

Melting Point 157°C to 161°C
Quantity 10 g
UN Number UN2811
Solubility Information Insoluble in water.
Formula Weight 172.57
Percent Purity 95%
Chemical Name or Material 3-Chloro-4-nitroaniline
Hazard Category H302+H332-H311-H315-H319
Hazard Statement GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only