accessibility menu, dialog, popup

UserName

NaH3COO

CAS RN 822-16-2

IUPAC Name: sodium octadecanoate
Synonyms: sodium stearate
Molecular Weight (g/mol): 306.47
Molecular Formula: C18H35NaO2
InChi Key: RYYKJJJTJZKILX-UHFFFAOYSA-M
SMILES: [Na+].CCCCCCCCCCCCCCCCCC([O-])=O
Molecular Weight (g/mol) 
  • (4)
  • (3)
Physical Form 
  • (1)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (3)
  • (4)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)