missing translation for 'onlineSavingsMsg'
Learn More

Sodium Stearate 97.0+%, TCI America™

Catalog No. S008125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S008125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S008125G Supplier TCI America Supplier No. S008125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Anionic surfactant used as a chemical reagent.

Specifications

Melting Point 220°C
Color White
CAS 822-16-2
Physical Form Crystalline Powder
Quantity 25 g
Molecular Formula C18H35NaO2
MDL Number MFCD00036404
UN Number 3077
Synonym sodium stearate, sodium octadecanoate, octadecanoic acid, sodium salt, stearates, stearic acid, sodium salt, prodhygine, flexichem b, stearic acid sodium salt, bonderlube 235, unii-qu7e2xa9tg
InChI Key RYYKJJJTJZKILX-UHFFFAOYSA-M
SMILES CCCCCCCCCCCCCCCCCC(=O)[O-].[Na+]
IUPAC Name sodium;octadecanoate
Molecular Weight (g/mol) 306.466
PubChem CID 2724691
Formula Weight 306.47
Percent Purity ≥97.0% (T)
Chemical Name or Material Sodium Stearate
Show More Show Less
EINECSNumber (2)-0611&(7)-0973
RTECSNumber WI4725000
TSCA Yes