missing translation for 'onlineSavingsMsg'
Learn More

Sodium Stearate, Powder, NF, 90%, Spectrum™ Chemical

Catalog No. 18606800
Change view
Click to view available options
Quantity:
2.5 kg
12 kg
45 kg
Packaging:
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606800 45 kg Poly Pail
18606798 2.5 kg Poly Pail
18606799 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18606800 Supplier Spectrum Chemical Mfg Cor Supplier No. S144745KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

S1447, 822-16-2, C18H35NaO2

Spectrum™ Chemical Sodium Stearate, Powder, NF is used in the pharmaceutical industry as a surfactant to aid the solubility of hydrophobic compounds. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 822-16-2
Molecular Formula C18H35NaO2
Molecular Weight (g/mol) 306.47
InChI Key RYYKJJJTJZKILX-UHFFFAOYSA-M
IUPAC Name sodium octadecanoate
SMILES [Na+].CCCCCCCCCCCCCCCCCC([O-])=O

Specifications

CAS Max % 1
Physical Form Powder, Needles or Crystals
Quantity 45 kg
Percent Purity 90%
Grade NF
Loss on Drying 0.05
Packaging Poly Pail
Chemical Name or Material Sodium Stearate, Powder