accessibility menu, dialog, popup

UserName

H3CCH2

CAS RN 821-95-4

IUPAC Name: undec-1-ene
Synonyms: 1-undecene undecene (mixture of isomers)
Molecular Weight (g/mol): 154.3
Molecular Formula: C11H22
InChi Key: DCTOHCCUXLBQMS-UHFFFAOYSA-N
SMILES: CCCCCCCCCC=C
Boiling Point 
  • (3)
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)