missing translation for 'onlineSavingsMsg'
Learn More

1-Undecene 99.5+%, TCI America™

Catalog No. S03415ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S03415ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S03415ML Supplier TCI America Supplier No. S03415ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 821-95-4
Molecular Formula C11H22
Molecular Weight (g/mol) 154.297
MDL Number MFCD00008956
InChI Key DCTOHCCUXLBQMS-UHFFFAOYSA-N
Synonym 1-undecene, undecene, n-1-undecene, alpha-undecene, alpha-undecylene, 1-hendecene, alpha-nonylethylene, undecene-1, undecene petroleum, ccris 5720
PubChem CID 13190
ChEBI CHEBI:77444
IUPAC Name undec-1-ene
SMILES CCCCCCCCCC=C

Specifications

Color Colorless
Boiling Point 193°C
Quantity 5 mL
Formula Weight 154.30
Percent Purity ≥99.5% (GC)
Physical Form Liquid
Chemical Name or Material 1-Undecene [Standard Material for GC]
EINECSNumber (2)-0027
TSCA Yes