missing translation for 'onlineSavingsMsg'
Learn More

1-Undecene 98.0+%, TCI America™

Catalog No. U005225ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U005225ML 25 mL
U0052100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. U005225ML Supplier TCI America Supplier No. U005225ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 821-95-4
Molecular Formula C11H22
Molecular Weight (g/mol) 154.297
MDL Number MFCD00008956
InChI Key DCTOHCCUXLBQMS-UHFFFAOYSA-N
Synonym 1-undecene, undecene, n-1-undecene, alpha-undecene, alpha-undecylene, 1-hendecene, alpha-nonylethylene, undecene-1, undecene petroleum, ccris 5720
PubChem CID 13190
ChEBI CHEBI:77444
IUPAC Name undec-1-ene
SMILES CCCCCCCCCC=C

Specifications

Melting Point -49°C
Color Yellow
Boiling Point 190°C
Quantity 25 mL
Formula Weight 154.30
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Undecene
EINECSNumber (2)-0027
TSCA Yes