accessibility menu, dialog, popup

UserName

CH3OPClOOH3C

CAS RN 814-49-3

IUPAC Name: diethyl phosphorochloridate
Synonyms: diethyl chlorophosphate
Molecular Weight (g/mol): 172.54
Molecular Formula: C4H10ClO3P
InChi Key: LGTLXDJOAJDFLR-UHFFFAOYSA-N
SMILES: CCOP(Cl)(=O)OCC
Boiling Point 
  • (2)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (1)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (4)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)