accessibility menu, dialog, popup

UserName

H3COBrOH

CAS RN 80866-82-6

IUPAC Name: (5-bromo-2-methoxyphenyl)methanol
Synonyms: (5-bromo-2-methoxyphenyl)methanol
Molecular Weight (g/mol): 217.06
Molecular Formula: C8H9BrO2
InChi Key: HAOOGRLZVQWDBY-UHFFFAOYSA-N
SMILES: COC1=CC=C(Br)C=C1CO
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results