CAS RN 8001-30-7
CAS RN 8001-30-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Corn oil
CAS: 8001-30-7 MDL Number: MFCD00130865
2
Corn Oil, MP Biomedicals
CAS: 8001-30-7
3
Corn Oil, NF, Spectrum™ Chemical
CAS: 8001-30-7