missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corn Oil, NF, Spectrum™ Chemical
Description
Spectrum™ Chemical Corn Oil, NF its uses include as a solvent vehicle for topical preparations and as a glidant excipient for oral dosage tablets and capsules. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 100% |
| Packaging | Amber Glass Bottle |
| Quantity | 25 mL |
| Grade | NF |