accessibility menu, dialog, popup

UserName

H3COSOOCH3

CAS RN 80-48-8

IUPAC Name: methyl 4-methylbenzene-1-sulfonate
Synonyms: methyl 4-methylbenzenesulfonate
Molecular Weight (g/mol): 186.23
Molecular Formula: C8H10O3S
InChi Key: VUQUOGPMUUJORT-UHFFFAOYSA-N
SMILES: COS(=O)(=O)C1=CC=C(C)C=C1
Boiling Point 
  • (2)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (4)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)