missing translation for 'onlineSavingsMsg'
Learn More

Methyl p-Toluenesulfonate 98.0+%, TCI America™

Catalog No. T0269500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0269500G 500 g
T026925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0269500G Supplier TCI America Supplier No. T0269500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 80-48-8
Molecular Formula C8H10O3S
Molecular Weight (g/mol) 186.225
MDL Number MFCD00008417
InChI Key VUQUOGPMUUJORT-UHFFFAOYSA-N
Synonym methyl p-toluenesulfonate, methyl tosylate, methyl p-tosylate, methyl toluene-4-sulfonate, methyl-p-toluenesulfonate, p-toluenesulfonic acid, methyl ester, benzenesulfonic acid, 4-methyl-, methyl ester, methyl toluene-4-sulphonate, p-toluenesulfonic acid methyl ester, p-methylbenzenesulfonate methyl ester
PubChem CID 6645
IUPAC Name methyl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OC

Specifications

Color White-Yellow
Boiling Point 145°C
Quantity 500 g
UN Number 3261
Formula Weight 186.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Methyl p-Toluenesulfonate
EINECSNumber (3)-1896
RTECSNumber XT7000000
TSCA Yes