missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl p-toluenesulfonate, 97%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Acidity | 0.2% max. |
| Boiling Point | 292.0°C |
| Flash Point | >110°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 96% min. (GC) |
| Packaging | Glass bottle |
| Refractive Index | 1.5162 to 1.5182 |
| Linear Formula | CH3C6H4SO3CH3 |
| Quantity | 100g |
| Beilstein | 11, 99 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. Harmful if swallowed. May cause an allergic skin reaction. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. IF ON SKIN ( |
| EINECSNumber | 201-283-5 |
| RTECSNumber | XT7000000 |
| TSCA | TSCA |
RUO – Research Use Only