accessibility menu, dialog, popup

UserName

CAS RN 79955-27-4

IUPAC Name: 3',6'-bis(acetyloxy)-3-oxo-3H-spiro[2-benzofuran-1,9'-xanthene]-5-carboxylic acid
Synonyms: 3',6'-bis(acetyloxy)-3-oxospiro[2-benzofuran-1,9'-xanthene]-5-carboxylic acid
Molecular Weight (g/mol): 460.39
Molecular Formula: C25H16O9
InChi Key: WPUZGNPQMIWOHE-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC=C2C(OC3=CC(OC(C)=O)=CC=C3C22OC(=O)C3=CC(=CC=C23)C(O)=O)=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)