missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ 5-Carboxyfluorescein Diacetate, Calbiochem™,

Catalog No. 21627550MG
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21627550MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21627550MG Supplier MilliporeSigma™ Supplier No. 21627550MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Non-fluorescent, hydrophobic agent that permeates intact cell membranes. In the cell, it is enzymatically hydrolyzed to a hydrophilic agent, 5-Carboxyfluorescein that exhibits strong fluorescence

Chemical Identifiers

CAS 79955-27-4
Molecular Formula C25H16O9
Molecular Weight (g/mol) 460.394
InChI Key WPUZGNPQMIWOHE-UHFFFAOYSA-N
Synonym 5-carboxyfluorescein diacetate, 5-cfda, 3',6'-diacetoxy-3-oxo-3h-spiro isobenzofuran-1,9'-xanthene-5-carboxylic acid, 5-carboxy-di-o-acetylfluorescein, 3',6'-diacetyloxy-3-oxospiro 2-benzofuran-1,9'-xanthene-5-carboxylic acid, cfda-5, spiro isobenzofuran-1 3h ,9'-9h xanthene-5-carboxylicacid, 3',6'-bis acetyloxy-3-oxo, 3',6'-bis acetyloxy-3-oxospiro 2-benzofuran-1,9'-xanthene-5-carboxylic acid, 3',6'-bis acetyloxy-3-oxospiro isobenzofuran-1 3h ,9'-9h xanthene-5-carboxylic acid, 3',6'-diacetoxy-3-oxo-3h-spiro 2-benzofuran-1,9'-xanthene-5-carboxylic acid
PubChem CID 133314
IUPAC Name 3',6'-diacetyloxy-3-oxospiro[2-benzofuran-1,9'-xanthene]-5-carboxylic acid
SMILES CC(=O)OC1=CC2=C(C=C1)C3(C4=C(C=C(C=C4)C(=O)O)C(=O)O3)C5=C(O2)C=C(C=C5)OC(=O)C

Specifications

Color Yellow
Quantity 50 mg
Solubility Information Soluble in DSMO
Formula Weight 460.4
Percent Purity ≥95%
Grade HPLC
Physical Form Solid
Chemical Name or Material 5-Carboxyfluorescein Diacetate
Recommended Storage -20°C (-4°F)