accessibility menu, dialog, popup

UserName

CH3NNH3CCl

CAS RN 79917-88-7

IUPAC Name: 1,3-dimethyl-1H-imidazol-3-ium chloride
Synonyms: 1,3-dimethylimidazolium chloride
Molecular Weight (g/mol): 132.59
Molecular Formula: C5H9ClN2
InChi Key: IIJSFQFJZAEKHB-UHFFFAOYSA-M
SMILES: [Cl-].CN1C=C[N+](C)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results