missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethylimidazolium Chloride 98.0+%, TCI America™

Catalog No. D33415G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33415G 5 g
D334125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D33415G Supplier TCI America Supplier No. D33415G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 79917-88-7
Molecular Formula C5H9ClN2
Molecular Weight (g/mol) 132.591
InChI Key IIJSFQFJZAEKHB-UHFFFAOYSA-M
PubChem CID 11062432
IUPAC Name 1,3-dimethylimidazol-1-ium;chloride
SMILES CN1C=C[N+](=C1)C.[Cl-]

Specifications

Melting Point 125°C
Color White-Yellow
Quantity 5 g
Formula Weight 132.59
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Dimethylimidazolium Chloride